Learn More
Thermo Scientific™ Freezer and Refrigerator Shelves
Product Code. 10123692 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
23.3, 45.8, 51.1, 78.8 cu. ft. Units
29.2 cu. ft. Units
4.7 cu. ft., 11.5 cu. ft. Units
51.1 cu. ft. Units
Type:
Open Wire Shelves
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Type Unit Size
10123692 23.3, 45.8, 51.1, 78.8 cu. ft. Units Open Wire Shelves Each
11762318 29.2 cu. ft. Units Open Wire Shelves Each
16655624 29.2 cu. ft. Units Open Wire Shelves Each
11747754 29.2 cu. ft. Units Open Wire Shelves Each
11749863 51.1 cu. ft. Units Open Wire Shelves Each
11752318 4.7 cu. ft., 11.5 cu. ft. Units Open Wire Shelves Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10123692 Supplier Thermo Scientific™ Supplier No. 6696
  • ONLINE EXCLUSIVE PRICE
    147.84€
    136.00€ / Pack of 1
    Save 11.84€
    8% Off
View all promotions
 Restricted Item
Estimated Shipment: 04-01-2027
Maximize lab storage with Thermo Scientific™ Freezer and Refrigerator Shelves, designed for use as replacement shelves in high-performance laboratory refrigerators, freezers and ULTs.
Maximize lab storage with Thermo Scientific™ Freezer and Refrigerator Shelves, designed for use as replacement shelves in high-performance laboratory refrigerators, freezers and ULTs.
  • Epoxy-coated open-wire shelves
  • Full- and half-depth sizes
  • Undercounter units use full-depth shelves only
TRUSTED_SUSTAINABILITY
Specifications
Type Open Wire Shelves
Exterior Finish Epoxy-coated Steel
For Use With (Equipment) 23.3, 45.8, 51.1, 78.8 cu. ft. Units
Warranty Warranty varies by country, please contact us for details
Holds General Freezer Items