Learn More
Eppendorf™ Freezer Rack: Innova™ U360, U535, U725/-G, CryoCube™ F740 series (3-comp., Std),
Product Code. 15713235
Change view
Click to view available options
Product Type:
Freezer Rack
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
15713235 Freezer Rack Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15713235 Supplier Eppendorf™ Supplier No. 6001011210
In Stock
Add to basket
Add to basket
Comprehensive storage of samples in a ULT freezer is based on storage boxes. The boxes should be stored in dedicated freezer racks. These metal racks enable optimal use of space as well as easy relocation of sample boxes.
Freezer Rack: Innova™ U360, U535, U725, U725-G, CryoCube™ F740 series (3-compartment, Standard), 2 in/53 mm, side access, stainless steel- Rack for 53 mm/2 in boxes- Max. box size [mm]: 136 x 136- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 569 x 139 x 389 (22.4 x 5.5 x 15.3 in)- Max. box number per rack: 28 (4 x 7)- Stainless steel- For ULT freezer: Innova U360 (max. 9 racks), Innova U535 (max. 12 racks), Innova U725 and U725-G (max. 18 racks), CryoCube F740 series (3-compartment version; max. 18 racks)- For ULT freezer: Innova U360 (max. 252 boxes), Innova U535 (max. 336 boxes), Innova U725 and U725-G (max. 504 boxes), CryoCube F740 series (3-compartment version; max. 504 boxes)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Freezer Rack