Learn More
Tenak™ Freezer Rack for Goblets
Freezer Rack designed for use in cryogenic LN2 freezers.
96.60€ - 107.00€
Specifications
Includes | Label Holder, Grip Lifter |
---|---|
Material | Stainless Steel |
For Use With (Equipment) | Standard 65 mm goblets with a height of 125 mm |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
12408410
|
Tenak™
TE21928 |
Goblets (5) |
107.00€
Each |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||||
12468400
|
Tenak™
TE21922 |
Goblets (2) |
96.60€
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
12488400
|
Tenak™
TE21924 |
Goblets (3) |
96.60€
Each |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||||
12498400
|
Tenak™
TE21926 |
Goblets (4) |
102.00€
Each |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||||
Description
- Goblet holds between 50 and 70 straws
- Two springs per shelf secure the goblet from slipping out
- Includes label holder for easy identification of the samples
- Includes a grip lifter, which makes it easier to grab the rack when wearing cryo gloves
- Constructed of stainless steel
Specifications
Label Holder, Grip Lifter | |
Stainless Steel | |
Standard 65 mm goblets with a height of 125 mm |