Learn More
Tenak™ Freezer Rack for Goblets
Freezer Rack designed for use in cryogenic LN2 freezers.
Brand: Tenak™ TE21926
Additional Details : Weight : 1.92000kg
Description
- Goblet holds between 50 and 70 straws
- Two springs per shelf secure the goblet from slipping out
- Includes label holder for easy identification of the samples
- Includes a grip lifter, which makes it easier to grab the rack when wearing cryo gloves
- Constructed of stainless steel
Specifications
Label Holder, Grip Lifter | |
Stainless Steel | |
70 x 70 x 575 mm |
Goblets (4) | |
Standard 65 mm goblets with a height of 125 mm |