Learn More
Karl Hecht™ Freezer-Fridge Thermometer
Click to view available options
Temperature Range (Metric):
-40°C to +50°C
Unit Size:
Each
Description
- Provided with a cylindrical plastic case
- Designed for hanging
- Colored filling
- Temperature range from −40° to +50°C
- Approx. 21cm long
Specifications
Specifications
| Includes | Plastic case |
| Length (Metric) | ∼21 cm |
| Temperature Range (Metric) | -40°C to +50°C |
| For Use With (Equipment) | Freezers, fridges |