Learn More
Thermo Scientific™ Storage Boxes
Product Code. 11976924 Shop All Thermo Scientific Products
Change view
Click to view available options
Holds:
100 (1, 1.5, 2 mL Cryo Tubes)
100 (3 and 4 mL Cryo Tubes)
100 Cryo Tubes
81 (1, 1.5, 2 mL Cryo Tubes)
81 Cryo Tubes
Cryo Tubes
Dimensions (L x W x H):
5 x 5 x 2 in.
5 x 5 x 2 in. (13 x 13 x 5 cm)
5 x 5 x 3 in.
5.3 x 5.3 x 2 in.
5.3 x 5.3 x 3.1 in.
Unit Size:
Each
Pack of 12
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Dimensions (L x W x H) unitSize
11976924 Cryo Tubes 5 x 5 x 3 in. Pack of 12
10696335 81 Cryo Tubes 5 x 5 x 2 in. (13 x 13 x 5 cm) Each
11814560 81 Cryo Tubes 5 x 5 x 3 in. Each
11349341 100 Cryo Tubes 5 x 5 x 2 in. Each
13487248 100 (3 and 4 mL Cryo Tubes) 5.3 x 5.3 x 3.1 in. Each
13467248 81 (1, 1.5, 2 mL Cryo Tubes) 5.3 x 5.3 x 2 in. Each
13457238 100 (1, 1.5, 2 mL Cryo Tubes) 5.3 x 5.3 x 2 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11976924 Supplier Thermo Scientific™ Supplier No. 5956
  • ONLINE EXCLUSIVE PRICE
    53.35€
    52.30€ / Pack of 12
    Save 1.05€
    1% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Store a variety of vials and tubes with these fiberboard storage boxes, for use in ultra-low temperature freezers.
Store a variety of vials and tubes with Thermo Scientific™ Storage Boxes, for use in ultra-low temperature freezers.
  • Sturdy, water-repellent boxes with covers for sample storage
  • Available in 2 and 3 in. high (5.1 and 7.6 cm) with 5.25 x 5.25 in. (13.3 x 13.3 cm) square bases
  • Box dividers included excluding 5954 and 5956
TRUSTED_SUSTAINABILITY
Specifications
Color White
Material Fiberboard
Shape Square
Type Cryo Storage Box
For Use With (Equipment) Thermo Scientific Ultra-Low Temperature Freezers
Holds Cryo Tubes
Includes Does not include dividers
Dimensions (L x W x H) 5 x 5 x 3 in.
Length (English) 5 in.
Length (Metric) 12.7 cm
Width (English) 5 in.
Width (Metric) 12.7 cm
Height (English) 3 in.
Height (Metric) 8 cm
Weight (Metric) 0.66 kg