Learn More
Thermo Scientific™ Frames and Caps for Immuno Standard Modules
BIOPHARMA
Product Code. 10524811
Change view
Click to view available options
Color:
Clear
White
Sterility:
Non-sterile
Sterile
Product Type:
Frame
Strip Cap
Unit Size:
Pack of 120
Pack of 180
Pack of 60
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Sterility Product Type Unit Size
10524811 White Non-sterile Frame Pack of 60
10224551 Clear Non-sterile Strip Cap Pack of 180
10684022 Clear Sterile Strip Cap Pack of 120
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10524811 Supplier Thermo Scientific™ Supplier No. 460348
  • ONLINE EXCLUSIVE PRICE
    91.60€
    89.99€ / Pack of 60
    Save 1.61€
    1% Off
Estimated Shipment: 09-10-2026
Add to basket
Add to basket
Bulk Quote Request
Easily insert and remove strip plates with solid modules by utilizing these frames and caps for immuno standard modules.
Easily insert and remove strip plates with solid modules by utilizing Thermo Scientific™ Frames and Caps for Immuno Standard Modules.
  • Polyethylene, stackable frame is marked for lid orientation
TRUSTED_SUSTAINABILITY
Specifications
Color White
Product Type Frame
For Use With (Equipment) Nunc Modules
Material Polyethylene
Sterility Non-sterile
No. per Case 60
No. per Pack 5
Plate Compatibility Immuno Standard Modules
Quantity 60/Cs.