missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FOSL1 (Texas Red/FITC) Split FISH Probe
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
1475.00€ - 2430.00€
Specifications
Content And Storage | Store at 4°C in the dark. |
---|---|
Description | Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique. |
Format | Tube |
Label or Dye | FITC, Texas Red |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16093465
|
Abnova™
FS0039.200UL |
200 μL |
2430.00€
200µL |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
16083465
|
Abnova™
FS0039.100UL |
100 μL |
1475.00€
100µL |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
- Dual colored probes for fast, sensitive, and specific detection
- Work on metaphase spread, paraffin embedded, and frozen tissue
- Identify gene amplification, loss, and translocation
- High signal to noise ratio
- Low cross reactivity
Specifications
Store at 4°C in the dark. | |
Tube | |
Fluorescent In Situ Hybridization (Cell) | |
11q13/11q13 | |
FOSL1 Split FISH Probe | |
Human |
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique. | |
FITC, Texas Red | |
DAPI Counterstain (1500 ng/mL ) 250 uL | |
Approx. 460kb/720kb | |
Genomic DNA | |
Liquid |