missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fosetyl-aluminum

Product Code. p-7214820 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 17201196

Brand: Thermo Scientific Chemicals 467280250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39148-24-8 | C6H15AlO9P3, C6H18AlO9P3 | 351.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39148-24-8
Molecular Formula C6H15AlO9P3

Specifications

Quantity 25 g
Solubility Solubility aq. soln.: 120 g/L (20°C). Other solubilities: practically insoluble in acetonitrile, propylene, glycol
Melting Point >300.0°C
Formula Weight 354.11
InChI Key OIPMQULDKWSNGX-UHFFFAOYSA-N
SMILES [Al+3].CCOP(=O)=O.CCOP(=O)=O.CCOP(=O)=O
IUPAC Name aluminium(3+) tris((phosphooxy)ethane)
Molecular Weight (g/mol) 351.08
Chemical Name or Material Fosetyl-Aluminum

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.