Learn More
Poulten Graf™ Fortuna™ Polyfix Natural Glass Dispensers
Product Code. p-8001043
Click to view available options
Volume (Metric):
10 to 50 mL
2 to 10 mL
Unit Size:
Each
Product Code. 10248190
Brand: Poulten Graf™ 101 07038
Only null left
Add to basket
Add to basket
For dispensing of all chemicals - including solvents - that do not attack polypropylene and don't crystallize. Poulten Graf™ Fortuna™ Polyfix Natural Glass Dispensers come with a glass piston.
For dispensing of all chemicals - including solvents - that do not attack polypropylene and don't crystallize. Poulten Graf™ Fortuna™ Polyfix Natural Glass Dispensers come with a glass piston.
  • Visible dispensing procedure
  • Easy to use volume setting system
  • Valve block made of polypropylene with FIX adapter
Specifications
Accuracy 0.60%
Product Type Bottletop Dispenser
Color Natural
Material Glass
Volume (Metric) 2 to 10 mL
Increments 0.2 mL