Learn More
Poulten Graf™ Fortuna™ Optima™ All Glass Luer-Lock Syringe
Able to be autoclaved at 134°C. Poulten Graf™ Fortuna™ Optima™ All Glass Luer-Lock Syringes come with an interchangeable piston.
9.90€ - 55.50€
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | CE EN 46001 |
Material | Borosilicate Glass |
Description | Amber graduation |
Product Type | All Glass Luer-lock Syringe |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10262151
|
Poulten Graf™
7 14021 |
1 mL |
9.90€
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
10252111
|
Poulten Graf™
7 14037 |
10 mL |
12.95€
Each |
In Stock
Log in to see stock. |
|||||
10383851
|
Poulten Graf™
7 14027 |
2 mL |
9.90€
Each |
In Stock
Log in to see stock. |
|||||
10353951
|
Poulten Graf™
7 14041 |
20 mL |
19.25€
Each |
In Stock
Log in to see stock. |
|||||
10249160
|
Poulten Graf™
7 14044 |
30 mL |
23.60€
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
10507941
|
Poulten Graf™
7 14033 |
5 mL |
10.65€
Each |
In Stock
Log in to see stock. |
|||||
10538321
|
Poulten Graf™
7 14045 |
50 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Description
- Complies with CE EN 46001 standards
- Constructed from Durobax™ glass
- With permanent amber graduation
- Acid and alkali resistant
Specifications
Autoclavable | |
Borosilicate Glass | |
All Glass Luer-lock Syringe | |
Luer Lock |
CE EN 46001 | |
Amber graduation | |
1/Ea. | |
Yes |