Learn More
Poulten Graf™ Fortuna™ Optima™ All Glass Luer-Lock Syringe
Able to be autoclaved at 134°C. Poulten Graf™ Fortuna™ Optima™ All Glass Luer-Lock Syringes come with an interchangeable piston.
Brand: Poulten Graf™ 7 14021
Additional Details : Weight : 0.02000kg
Description
- Complies with CE EN 46001 standards
- Constructed from Durobax™ glass
- With permanent amber graduation
- Acid and alkali resistant
Specifications
Autoclavable | |
Borosilicate Glass | |
All Glass Luer-lock Syringe | |
1/Ea. | |
Luer Lock |
CE EN 46001 | |
Amber graduation | |
0.05 mL | |
1 mL | |
Yes |