Learn More
Poulten Graf™ Fortuna™ Optifix™ Safety Dispenser
Product Code. 10549491
Click to view available options
Volume (Metric):
0.4 to 2 mL
1 to 5 mL
10 to 50 mL
2 to 10 mL
2.0 to 10 mL
20 to 100 mL
6 to 30 mL
Includes:
PTFE-coated Piston, Stopcock and Ventilator Valve
PTFE-encased Piston, Sealing Lip
Strongly PTFE-encased Special Piston
Unit Size:
Each
Product Code. 10549491
Brand: Poulten Graf™ 101 10744
For dispensing of strong acids and aggressive chemicals (no HF). Poulten Graf™ Fortuna™ Optifix™ Safety Dispensers have a dispensing cylinder made of glass and a PTFE-encased piston with sealing lip.
For dispensing of strong acids and aggressive chemicals (no HF). Poulten Graf™ Fortuna™ Optifix™ Safety Dispensers have a dispensing cylinder made of glass and a PTFE-encased piston with sealing lip.
  • Manufactured according to DIN EN ISO 9001:2008 using high quality materials
  • With certificate, individually numbered
  • Glass dispensing cylinder is coated with plastic to provide breakage protection
  • Ergonomic finger protection cap made of PP with volume setting system
  • Graduation on the scale is lasered and easy to read
  • Fast and easy volume setting with additional fine adjustment
Specifications
Product Type Safety Dispenser
Volume (Metric) 6 to 30 mL
Includes PTFE-encased Piston, Sealing Lip