missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formic Acid-OD (95% w/w in D2O), CDN
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Formic Acid-d |
| CAS | 917-71-5 |
| Solvent or Matrix | D2O |
| Molecular Formula | C D H O2 |
| Quantity | 5 g |
| Synonym | Formic-d acid (6 CI, 7 CI, 8 CI, 9 CI), Deuterioformic acid, Formic-D acid, Formic acid-d1, Formic-d1 acid |
| InChI Formula | InChI=1 S/CH2O2/c2-1-3/h1H,(H,2,3)/i1D |
| SMILES | [2 H]C(=O)O |
| IUPAC Name | deuterioformic acid |
| Molecular Weight (g/mol) | 47.0315 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction