missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde (37% w/w aq. soln., Stabilized 7-8% of Methanol), TRC

Product Code. 30007838
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
Unit Size:
1 L
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007838 1 L 1 L
30004982 250 mL 250mL
30004825 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007838 Supplier Toronto Research Chemicals Supplier No. TRCF6913501L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-00-0
Formula Weight 30.0106
InChI Formula InChI=1S/CH2O/c1-2/h1H2
IUPAC Name formaldehyde
Molecular Formula C H2 O
Molecular Weight (g/mol) 30.03
SMILES C=O
Synonym Formaldehyde, Methanal, BFV, F-gen, FM 282, FS 850A, Fannoform, Floguard 1015, Fordor, Formaldehyde-12C, Formalin, Formalin LM, Formalin Taisei, Formalith, Formic aldehyde, Formol, Fyde, Lysoform, Methaldehyde, Methyl aldehyde, Methylene oxide, Morbicid, NSC 298885, Optilyse, Oxomethane, Oxymethylene, Paraform, Superlysoform
Concentration or Composition (by Analyte or Components) Formaldehyde 37%
Chemical Name or Material Formaldehyde

Specifications

Quantity 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.