Learn More
Rogo Sampaic™ Forceps
Click to view available options
Length (Metric):
12 cm
14 cm
missing translation for 'unitSize'
Pack of 10
Specifications
Specifications
| Product Type | Forceps |
| Material | Stainless Steel |
| For Use With (Application) | For Tissue |
| Length (Metric) | 14 cm |