Learn More
Cole-Parmer Cleanroom Hose Barb Fittings, Y Union, High-Density Polyethylene
Shop All Cole-Parmer Products
Click to view available options
Diameter (English) Inner:
1/2 in.
3/16 in.
Unit Size:
Pack of 10
Description
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Quick and easy assembly without tools. Single barb design tightly grips the tubing. Meets FDA requirements 21 CFR 177.1520 (when used unmodified for the manufacture of food contact articles) and RoHS compliant
Specifications
Specifications
| Certifications/Compliance | FDA;Non-DEHP;Non-Phthalate;RoHS |
| Diameter (English) Inner | 1/2 in. |
| Packaging Quantity | 10/Pk. |
| Outlet Port | Hose Barb Y |
| Shape | Y |
| Color | White |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.