Learn More
Hartmann Larochette™ Foliodress™ Non-woven Surgical Face Masks
Click to view available options
Color:
Blue
Green
Unit Size:
Pack of 50
Description
- High-quality surgical face masks according to EN 14683:2014 type II or type II-R.
- Hypoallergenic non-woven materials, free from natural rubber latex.
- High filtration performance and comfortable breathing resistance
- Long integrated nose bar.
- Fragrance free.
- Durable.
Specifications
Specifications
| Product Type | Surgical Face Mask |
| Color | Blue |
| Sterility | Non-sterile |
| Certifications/Compliance | EN 14683:2014 type II or type II-R |
| Construction | Non-woven |
| Disposable | Single-use |
| Filter Efficiency | Bacterial |
| For Use With (Application) | For single-use in all surgical areas, as well as on isolation wards and intensive care departments. |