Learn More
Brady™ Foam Mounting Tape
Product Code. 16531350 Shop All Brady Products
Change view
Click to view available options
Dimensions (L x W):
32.92 m x 25 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
16531350 32.92 m x 25 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16531350 Supplier Brady™ Supplier No. 019811
  • ONLINE EXCLUSIVE PRICE
    69.95€
    68.79€ / Pack of 1
    Save 1.16€
    1% Off
View all promotions
Estimated Shipment: 09-11-2026
Indoor/outdoor B-156 double-faced foam tape is coated on both sides with high-tack, high-shear synthetic rubber adhesive and bonds well to even your roughest irregular surfaces.
  • Minimum application temperature is 10°C and service range is 10°C to 82°C
  • Permanent Synthetic Rubber Adhesive
Specifications
Product Type Self-Adhesive Mounting Tape
Color White
Material Polyethylene
Dimensions (L x W) 32.92 m x 25 mm