Learn More
Sartorius™ FN 4 Chromatography Paper
Product Code. 10138581 Shop All Sartorius Products
Change view
Click to view available options
Length (Metric):
580 mm
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10138581 580 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10138581 Supplier Sartorius™ Supplier No. FT2504580600N
 Restricted Item
Estimated Shipment: 17-11-2026
Specifications
Format Sheet
Grade FN 4
Material Cotton Linters
Width (Metric) 600 mm
Length (Metric) 580 mm
Technique Chromatography