missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Fmk
GREENER_CHOICE

Product Code. 18799666
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 18799666

Brand: Tocris Bioscience™ 4690/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Irreversible ribosomal S6 kinase 1/2 inhibitor

Fmk is a potent irreversible RSK1/2 inhibitor (IC50 = 15 nM). Inactivates the C-terminal auto-kinase domain activity. Suppresses RSK2 kinase activity in RSK2-expressing HNSCC cell lines. Reduces HNSCC cell invasive ability. Displays no effect on EGF-stimulated phosphorylation of ERK1 or ERK2.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-[4-Amino-7-(3-hydroxypropyl)-5-(4-methylphenyl)-7H-pyrrolo[2,3-d]pyrimidin-6-yl]-2-fluoroethanone
CAS 821794-92-7
Quantity 10 mg
Molecular Formula C18H19FN4O2
Formula Weight 342.37
Purity 97%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.