Learn More
Greiner Bio-One FLUOTRAC™ 200 LoBase Flat-Bottom 1536-well Microplate
Description
Alphanumeric well coding
Specifications
Specifications
| Certifications/Compliance | Free of detectable DNase/RNase, human DNA and pyrogens |
| Format | Standard |
| Lid | Without Lid |
| Culture Area | 2.3 mm 2 |
| Array | 32 x 48 |
| Autoclavable | Not Autoclavable |
| Binding Type | Medium Binding |
| Color | Black |
| Material | Polystyrene |
| No. of Wells | 1536 |
| Show More |