Learn More
Greiner Bio-One FLUOTRAC™ 200 HiBase μClear™-Bottom 1536-well Microplate
GREENER_CHOICE
Product Code. 13037963
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 60
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
13037963 Clear Pack of 60
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13037963 Supplier Greiner Bio-One Supplier No. 782096
Only null left
Add to basket
Add to basket
Free of detectable DNase, RNase, human DNA and pyrogens
Alphanumeric well coding
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Free of detectable DNase/RNase, human DNA and pyrogens
Format Standard
Lid With Lid
Array 32 x 48
Autoclavable Not Autoclavable
Binding Type Medium Binding
Color Clear
Material Polystyrene
No. of Wells 1536
Volume (Metric) Working 3 to 10 μL
Sterility Non-sterile
Well Shape Rounded Square
Dnase Free Yes
Max. RCF 1500 x g (swinging-bucket rotor)
No. per Case 60
No. per Pack 15
RNase Free Yes
Temperature Range (Metric) -20°C to +60°C
Dimensions (L x W x H) 128 x 85 x 10 mm