Learn More
Greiner Bio-One FLUOTRAC™ 200 HiBase μClear™-Bottom 1536-well Microplate
Description
Alphanumeric well coding
Specifications
Specifications
| Certifications/Compliance | Free of detectable DNase/RNase, human DNA and pyrogens |
| Format | Standard |
| Lid | With Lid |
| Array | 32 x 48 |
| Autoclavable | Not Autoclavable |
| Binding Type | Medium Binding |
| Color | Clear |
| Material | Polystyrene |
| No. of Wells | 1536 |
| Volume (Metric) Working | 3 to 10 μL |
| Show More |