missing translation for 'onlineSavingsMsg'
Learn More

Fluorometholone Acetate, TRC

Product Code. 30395848
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395848 5 mg 5mg
30402737 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30395848 Supplier Toronto Research Chemicals Supplier No. TRCF5931505MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluorometholone Acetate
CAS 3801-06-7
Molecular Formula C24 H31 F O5
Quantity 5 mg
Synonym Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,17-dihydroxy-6α-methyl-, 17-acetate (6CI,7CI,8CI), (6α,11β)-17-(Acetyloxy)-9-fluoro-11-hydroxy-6-methylpregna-1,4-diene-3,20-dione, 6α-Methyl-9α-fluoro-17-acetoxy-21-deoxyprednisolone, 6α-Methyl-9α-fluoro-21-desoxyprednisolone-17-acetate, Eflone, Flarex, Fluorometholone acetate, NSC 47438, Oxylone acetate, U 17323
InChI Formula InChI=1S/C24H31FO5/c1-13-10-19-17-7-9-23(14(2)26,30-15(3)27)22(17,5)12-20(29)24(19,25)21(4)8-6-16(28)11-18(13)21/h6,8,11,13,17,19-20,29H,7,9-10,12H2,1-5H3/t13-,17-,19-,20-,21-,22-,23-,24-/m0/s1
SMILES C[C@H]1C[C@H]2[C@@H]3CC[C@](OC(=O)C)(C(=O)C)[C@@]3(C)C[C@H](O)[C@]2(F)[C@@]4(C)C=CC(=O)C=C14
IUPAC Name [(6S,8S,9R,10S,11S,13S,14S,17R)-17-acetyl-9-fluoro-11-hydroxy-6,10,13-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] acetate
Molecular Weight (g/mol) 418.5
Formula Weight 418.2156
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.