Learn More
Thermo Scientific™ Fluoro-Max Green and Red Dry Fluorescent Particles
Product Code. 11855052 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (Metric) Nominal:
5 μm
10 μm
15 μm
25 μm
30 μm
40 μm
50 μm
70 μm
80 μm
100 μm
120 μm
140 μm
160 μm
Quantity:
1 g
5 g
Color:
Green
Red
Unit Size:
1g
5g
38 product options available for selection
Product selection table with 38 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Nominal Quantity Color Unit Size
11855052 5 μm 1 g Green 1g
11865042 80 μm 5 g Green 5g
11885042 100 μm 5 g Green 5g
11805052 120 μm 5 g Green 5g
11815052 140 μm 1 g Green 1g
11825052 140 μm 5 g Green 5g
11835052 160 μm 1 g Green 1g
11845052 160 μm 5 g Green 5g
11865052 5 μm 5 g Green 5g
11875052 10 μm 1 g Green 1g
11885052 10 μm 5 g Green 5g
11895052 15 μm 1 g Green 1g
11805062 15 μm 5 g Green 5g
11825062 25 μm 5 g Green 5g
11835062 30 μm 1 g Green 1g
11845062 30 μm 5 g Green 5g
11855062 40 μm 1 g Green 1g
11865062 40 μm 5 g Green 5g
11875062 50 μm 1 g Green 1g
11885062 50 μm 5 g Green 5g
11895062 70 μm 1 g Green 1g
11805072 70 μm 5 g Green 5g
11835072 100 μm 1 g Red 1g
11845072 100 μm 5 g Red 5g
11865072 5 μm 5 g Red 5g
11885072 10 μm 5 g Red 5g
11805082 15 μm 5 g Red 5g
11815082 25 μm 1 g Red 1g
11825082 25 μm 5 g Red 5g
11835082 30 μm 1 g Red 1g
11845082 30 μm 5 g Red 5g
11895042 120 μm 1 g Green 1g
11875042 100 μm 1 g Green 1g
11855042 80 μm 1 g Green 1g
11895072 15 μm 1 g Red 1g
11875072 10 μm 1 g Red 1g
11815062 25 μm 1 g Green 1g
11855072 5 μm 1 g Red 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 38 options available.
38 options
Product Code. 11855052 Supplier Thermo Scientific™ Supplier No. 352
  • ONLINE EXCLUSIVE PRICE
    683.00€
    322.96€ / 1g
    Save 360.04€
    52% Off
 Restricted Item
Estimated Shipment: 15-12-2026
Improve sensitivity and detectability for analytical methods with fluorescent particles that feature bright, distinct colors.
Improve sensitivity and detectability for analytical methods with Thermo Scientific™ Fluoro-Max Green and Red Dry Fluorescent Particles, polymer particles that are internally dyed and feature bright, distinct colors.
Made of polystyrene, these Fluoro-Max Green and Red Dry Fluorescent Particles have a density of 1.05g/cm3 and a refractive index of 1.59 @ 589nm (25°C). They can be detected with an epifluorescence microscope, confocal microscope, fluorometer, fluorescence spectrophotometer, or an fluorescence activated cell sorter. They can also be detected using mineral light or black light (UV).
Note:
Spectral information is approximate and for reference only. The spectral properties of the dyes are dependent on their concentration and physical environment. The exact excitation and emission maxima may vary depending on the size and composition of the particles.
Recommended for:
  • Filter testing
  • Flow tracing
  • Biological research
TRUSTED_SUSTAINABILITY
Specifications
Additive May contain trace amount of dispersant
Content And Storage Store at room temperature in a dry area. Store upright and keep bottle tightly sealed. Respiratory protection from dust is recommended.
Contents Dry dyed polystyrene (DVB) microspheres
Color Green
Material Polystyrene Divinylbenzene (PS-DVB)
Product Type Fluorescent Particle
Dye Type Firefli Fluorescent Green (468/508nm)
Includes Package Insert Sheet
No. of Cells 1.45 x 1010/gram
Particle Density 1.05g/cm³
Quantity 1 g
Refractive Index 1.59 at 589nm (25°C)
Stability Product stable for a minimum of 24 months
Uniformity <18%
Diameter (Metric) Mean 5 μm
Diameter (Metric) Nominal 5 μm
Diameter (Metric) 5 μm