missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Contains a total of 9 fluorescent proteins ranging from 14 kDa to 200 kDa.
- It is designed as a molecular weight standard for the Quick Fluorescent Protein Buffer, pre-stained protein samples, and other fluorescent protein samples.
- Increased protein band intensity allows for better electrophoresis monitoring while performing assays.
- This product can also be used as a molecular weight marker for Western blot experiments, and the membrane-transferred fluorescent markers can be visualized on a UV or LED transilluminator.
Specifications
Specifications
| Detection Method | UV Light, LED Transilluminator |
| Number of Markers | 9 |
| Ready to Load | Yes |
| Sample Loading Volume | 5 μL |
| Size Range | 14 to 200 kDa |
| Form | Liquid |
| Quantity | 1 Vial |
| Storage Requirements | -20°C |
| Product Type | Fluorescent Protein Marker |
| Stain Type | Fluorescent |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?