missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Fluorescent Exosome Standards (Human Plasma)

Product Code. 18605710 Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 18605710

Brand: Novus Biologicals™ NBP311692

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly pure, lyophilized exosome standards with superior stability, optimal for multiple applications

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C in the dark. Avoid freeze-thaw cycles.
For Use With (Application) Flow Cytometry, Electron Microscopy
Excitation/Emission The excitation maximum of fluorescent exosome standards is 500 nm - 650 nm and emission maximum is 510 - 665 nm.
Format Human biofluids of healthy donors
Quantity 100 μg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.