missing translation for 'onlineSavingsMsg'
Learn More

Fluorescent Brightener 28, TRC

Product Code. 30396851
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396851 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30396851 Supplier Toronto Research Chemicals Supplier No. TRCF5977832.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluorescent Brightener 28
CAS 4193-55-9
Molecular Formula C40H42N12Na2O10S2
Quantity 2.5 g
Synonym 2,2'-Stilbenedisulfonic acid, 4,4'-bis[[4-anilino-6-[bis(2-hydroxyethyl)amino]-s-triazin-2-yl]amino]-, disodium salt (6CI,7CI), Benzenesulfonic acid, 2,2'-(1,2-ethenediyl)bis[5-[[4-[bis(2-hydroxyethyl)amino]-6-(phenylamino)-1,3,5-triazin-2-yl]amino]-, disodium salt (9CI), C.I. Fluorescent Brightening Agent 28, disodium salt (8CI), 4,4'-Bis[2-[bis(2-hydroxyethyl)amino]-4-anilino-s-triazin-6-ylamino]stilbene-2,2'-disulfonic acid disodium salt, 4,4'-Bis[4-anilino-6-bis(2-hydroxyethyl)amino-s-triazin-2-yl]amino-2,2'-stilbenedisulfonic acid disodium salt, BA (fluorescent brightener), Blancophor MS 31, Blancophor MS 72, Blancophor RG 96FS, Blankophor DS, Brightener BR 3, C.I. 40622, C.I. Fluorescent Brightener 28, Calcofluor White, Calcofluor White LRP, Calcofluor White M 2R, Calcofluor White ST, Cellufluor, Disodium 4,4'-bis(4-anilino-6-diethanolamino-s-triazin-2-ylamino)- 2,2'-stilbenesulfonate, Disodium 4,4'-bis[4-[bis(b-hydroxyethyl)amino]-6-phenylamino-1,3,5-triazin-2-ylamino]-2,2'-stilbenedisulfonate, Disodium 4,4'- bis[[4-anilino-6-[bis(2-hydroxyethyl)amino]-s-triazin-2-yl]amino]-2,2'-stilbenedisulfonate, Fluka 18909, Hiltamine Arctic White CWD, Kayaphor FB, Keyfluor White ST, Paper White BP, Phorwite AR, Photine C 410, Ranipal 4BM, Tinopal 5BMS, Tinopal HW, Tinopal LPW, Tinopal UNPA, Tinopal UNPA-GX, Tintophen MS 31, Tintophen MS 72, Uvitex 4BM
InChI Formula InChI=1S/C40H44N12O10S2.2Na/c53-21-17-51(18-22-54)39-47-35(41-29-7-3-1-4-8-29)45-37(49-39)43-31-15-13-27(33(25-31)63(57,58)59)11-12-28-14-16-32(26-34(28)64(60,61)62)44-38-46-36(42-30-9-5-2-6-10-30)48-40(50-38)52(19-23-55)20-24-56;;/h1-16,25-26,53-56H,17-24H2,(H,57,58,59)(H,60,61,62)(H2,41,43,45,47,49)(H2,42,44,46,48,50);;/q;2*+1/p-2/b12-11+;;
SMILES C1=CC=C(C=C1)NC2=NC(=NC(=N2)N(CCO)CCO)NC3=CC(=C(C=C3)/C=C/C4=C(C=C(C=C4)NC5=NC(=NC(=N5)NC6=CC=CC=C6)N(CCO)CCO)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;5-[[4-anilino-6-[bis(2-hydroxyethyl)amino]-1,3,5-triazin-2-yl]amino]-2-[(E)-2-[4-[[4-anilino-6-[bis(2-hydroxyethyl)amino]-1,3,5-triazin-2-yl]amino]-2-sulfonatophenyl]ethenyl]benzenesulfonate
Molecular Weight (g/mol) 960.95
Formula Weight 960.2384
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.