missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluorescent Brightener 28 (Technical Grade), TRC
Specifications
Specifications
| Chemical Name or Material | Fluorescent Brightener 28 |
| CAS | 4193-55-9 |
| Molecular Formula | C40H42N12Na2O10S2 |
| Quantity | 500 mg |
| Synonym | 2,2'-Stilbenedisulfonic acid, 4,4'-bis[[4-anilino-6-[bis(2-hydroxyethyl)amino]-s-triazin-2-yl]amino]-, disodium salt (6CI,7CI), Benzenesulfonic acid, 2,2'-(1,2-ethenediyl)bis[5-[[4-[bis(2-hydroxyethyl)amino]-6-(phenylamino)-1,3,5-triazin-2-yl]amino]-, disodium salt (9CI), C.I. Fluorescent Brightening Agent 28, disodium salt (8CI), 4,4'-Bis[2-[bis(2-hydroxyethyl)amino]-4-anilino-s-triazin-6-ylamino]stilbene-2,2'-disulfonic acid disodium salt, 4,4'-Bis[4-anilino-6-bis(2-hydroxyethyl)amino-s-triazin-2-yl]amino-2,2'-stilbenedisulfonic acid disodium salt, BA (fluorescent brightener), Blancophor MS 31, Blancophor MS 72, Blancophor RG 96FS, Blankophor DS, Brightener BR 3, C.I. 40622, C.I. Fluorescent Brightener 28, Calcofluor White, Calcofluor White LRP, Calcofluor White M 2R, Calcofluor White ST, Cellufluor, Disodium 4,4'-bis(4-anilino-6-diethanolamino-s-triazin-2-ylamino)- 2,2'-stilbenesulfonate, Disodium 4,4'-bis[4-[bis(b-hydroxyethyl)amino]-6-phenylamino-1,3,5-triazin-2-ylamino]-2,2'-stilbenedisulfonate, Disodium 4,4'- bis[[4-anilino-6-[bis(2-hydroxyethyl)amino]-s-triazin-2-yl]amino]-2,2'-stilbenedisulfonate, Fluka 18909, Hiltamine Arctic White CWD, Kayaphor FB, Keyfluor White ST, Paper White BP, Phorwite AR, Photine C 410, Ranipal 4BM, Tinopal 5BMS, Tinopal HW, Tinopal LPW, Tinopal UNPA, Tinopal UNPA-GX, Tintophen MS 31, Tintophen MS 72, Uvitex 4BM |
| InChI Formula | InChI=1S/C40H44N12O10S2.2Na/c53-21-17-51(18-22-54)39-47-35(41-29-7-3-1-4-8-29)45-37(49-39)43-31-15-13-27(33(25-31)63(57,58)59)11-12-28-14-16-32(26-34(28)64(60,61)62)44-38-46-36(42-30-9-5-2-6-10-30)48-40(50-38)52(19-23-55)20-24-56;;/h1-16,25-26,53-56H,17-24H2,(H,57,58,59)(H,60,61,62)(H2,41,43,45,47,49)(H2,42,44,46,48,50);;/q;2*+1/p-2/b12-11+;; |
| SMILES | C1=CC=C(C=C1)NC2=NC(=NC(=N2)N(CCO)CCO)NC3=CC(=C(C=C3)/C=C/C4=C(C=C(C=C4)NC5=NC(=NC(=N5)NC6=CC=CC=C6)N(CCO)CCO)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+] |
| IUPAC Name | disodium;5-[[4-anilino-6-[bis(2-hydroxyethyl)amino]-1,3,5-triazin-2-yl]amino]-2-[(E)-2-[4-[[4-anilino-6-[bis(2-hydroxyethyl)amino]-1,3,5-triazin-2-yl]amino]-2-sulfonatophenyl]ethenyl]benzenesulfonate |
| Molecular Weight (g/mol) | 960.95 |
| Formula Weight | 960.2384 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?