missing translation for 'onlineSavingsMsg'
Learn More

SCHOTT AG Lighting and Imaging™ Fluorescence Exitation Filter

Evenly illuminates the entire fiber bundle and can be used with all standard fiber optics for the respective light source. For use with cold light source KL2500LED.

Brand:  SCHOTT AG Lighting and Imaging™ 258313

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10553792

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

  • Insert filters are placed inside of the cold light source
  • Evenly illuminate the entire fiber bundle
  • Can be used with all standard fiber optics for the respective light source
  • Diameter: 28mm
Specifications

Specifications

Filter
Blue 450 nm
Cold Light Source KL2500LED
28 mm dia.
Fluorescent Blue
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers