Learn More
Fluorene-d10, CDN
Product Code. 30320904
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30320904
Brand: CDN CDND12980.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Fluorene D10
CAS 81103-79-9
Molecular Formula C13 D10
Quantity 0.5 g
Synonym 9 H-Fluorene-1,2,3,4,5,6,7,8,9,9-d10, Fluorene-d10, Perdeuterofluorene
InChI Formula InChI=1 S/C13H10/c1-3-7-12-10(5-1)9-11-6-2-4-8-13(11)12/h1-8 H,9H2/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9D2
SMILES [2 H]c1c([2 H])c([2 H])c2c(c1[2 H])c3c([2 H])c([2 H])c([2 H])c([2 H])c3C2([2 H])[2 H]
IUPAC Name 1,2,3,4,5,6,7,8,9,9-decadeuteriofluorene
Molecular Weight (g/mol) 176.2801
Formula Weight 176.141 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity