Learn More
Flumetsulam, TRC
Specifications
Specifications
| Chemical Name or Material | Flumetsulam |
| CAS | 98967-40-9 |
| Molecular Formula | C12 H9 F2 N5 O2 S |
| Quantity | 100 mg |
| Synonym | N-(2,6-Difluorophenyl)-5-methyl-[1,2,4]triazolo[1,5-a]pyrimidine-2-sulfonamide, Broadstrike, DE 498, Flumetsulam, Python, Python (pesticide), Scorpion, Scorpion (pesticide), XRD 498 |
| InChI Formula | InChI=1S/C12H9F2N5O2S/c1-7-5-6-19-11(15-7)16-12(17-19)22(20,21)18-10-8(13)3-2-4-9(10)14/h2-6,18H,1H3 |
| SMILES | Cc1ccn2nc(nc2n1)S(=O)(=O)Nc3c(F)cccc3F |
| IUPAC Name | N-(2,6-difluorophenyl)-5-methyl-[1,2,4]triazolo[1,5-a]pyrimidine-2-sulfonamide |
| Molecular Weight (g/mol) | 325.29 |
| Formula Weight | 325.0445 |