Learn More
Flumethasone Acetate, TRC
Product Code. 30005596
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30005596 50 mg 50mg
18002140 5 mg 5mg
18053720 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30005596 Supplier Toronto Research Chemicals Supplier No. TRCF45500550MG
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2823-42-9
Molecular Formula C24H30F2O6
Molecular Weight (g/mol) 452.49
Synonym Flumethasone 21-Acetate, 6α-Fluorodexamethasone 21-Acetate, 6α,9-Difluoro-11β,17,21-trihydroxy-16α-methyl-pregna-1,4-diene-3,20-dione 21-Acetate;
IUPAC Name [2-[(6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-difluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
SMILES C[C@@H]1C[C@H]2[C@@H]3C[C@@H](C4=CC(=O)C=C[C@@]4([C@]3([C@H](C[C@@]2([C@]1(C(=O)COC(=O)C)O)C)O)F)C)F
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C24H30F2O6/c1-12-7-15-16-9-18(25)17-8-14(28)5-6-21(17,3)23(16,26)19(29)10-22(15,4)24(12,31)20(30)11-32-13(2)27/h5-6,8,12,15-16,18-19,29,31H,7,9-11H2,1-4H3/t12-,15+,16+,18+,19+,21+,22+,23+,24+/m1/s1
Formula Weight 452.201
Chemical Name or Material Flumethasone Acetate