Learn More
Thermo Scientific™ Fluid Transfer Assembly, Y Set
Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
179.00€ - 437.00€
Product Code | Brand | Tubing Style | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Style | Price | Quantity & Availability | |||||
17800637
|
Thermo Scientific™
SH31108.01 |
3 x 6.35 mm (1/4 in.) ID x 11.11 mm (7/16 in.) wall; TPE 374; 2 x 0.51 m (20 in.) to 0.51 m (20 in.); MPC body-insert |
260.00€
Pack of 10 |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
17830687
|
Thermo Scientific™
SH31109.01 |
3 x 6.35 mm (1/4 in.) ID x 11.11 mm (7/16 in.) wall; TPE 374; 2 x 0.51 m (20 in.) to 0.51 m (20 in.); MPC in.sert-body |
263.00€
Pack of 10 |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
17810637
|
Thermo Scientific™
SH31107.01 |
3 x 9.52 mm (3/8 in.) ID x 15.88 mm (5/8 in.) wall; TPE 374; 2 x 0.51 m (20 in.) to 0.51 m (20 in.); MPC in.sert-body |
291.00€
Pack of 10 |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
17870627
|
Thermo Scientific™
SH31104.01 |
3 x 12.7 mm (1/2 in.) ID x 19.05 mm (3/4 in.) wall; TPE 374; 2 x 0.51 m (20 in.) to 0.51 m (20 in.); MPX body-insert |
391.00€
Pack of 10 |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
17820637
|
Thermo Scientific™
SH31110.01 |
3 x 3.22 mm (1/8 in.) ID x 6.35 mm (1/4 in.) wall; TPE 374; 2 x 0.51 m (20 in.) to 0.51 m (20 in.); Luer |
179.00€
Pack of 10 |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
17840687
|
Thermo Scientific™
SH31105.01 |
3 x 12.7 mm (1/2 in.) ID x 19.05 mm (3/4 in.) wall; TPE 374; 2 x 0.51 m (20 in.) to 0.51 m (20 in.); MPX in.sert-body |
394.00€
Pack of 10 |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
17890627
|
Thermo Scientific™
SH31106.01 |
3 x 9.52 mm (3/8 in.) ID x 15.88 mm (5/8 in.) wall; TPE 374; 2 x 0.51 m (20 in.) to 0.51 m (20 in.); MPC body-insert |
437.00€
Pack of 10 |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
Description
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process.- One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
- Multiple manufacturing sites ensure security in product supply
- Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
- Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Specifications
Y Set |