Learn More
Thermo Scientific™ Fluid Transfer Assembly, Pump Set
Shop All Thermo Scientific Products
Click to view available options
Tubing Size:
10 in. TPE 374, 19.7 in. Pharmed BPT
Connector Types:
Luer
MPC
MPX
Dimensions:
1/2 in. ID x 3/4 in. OD
1/4 in. ID x 3/8 in. OD
1/4 in. ID x 7/16 in. OD
1/8 in. ID x 1/4 in. OD
3/8 in. ID x 5/8 in. OD
Unit Size:
Pack of 10
Description
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process.
- One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
- Multiple manufacturing sites ensure security in product supply
- Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
- Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Specifications
Specifications
| Dimensions | 1/4 in. ID x 7/16 in. OD |
| Tubing Style | 6.35 mm (1/4 in.) ID x 11.11 mm (7/16 in.) OD; 0.25 m (10 in.) TPE 374 to 0.5 m (19.7 in.) Pharmed BPT to 0.25 m (10 in.); TPE 374; MPC |
| Connector Types | MPC |
| Product Type | Pump Set |
| Tubing Size | 10 in. TPE 374, 19.7 in. Pharmed BPT |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering