Learn More
BD™ Fluid A, Double Wrapped
Product Code. 15857731
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15857731 100 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15857731 Supplier BD™ Supplier No. 257603
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Packaged in two Stericlin™ bags before autoclaving for a bottle exterior free from microbial contaminants. BD™ Fluid A, Double Wrapped is a diluting and rinsing fluid for sterility tests of pharmaceutical products according to the membrane filtration method.
Fluid A is a diluting and rinsing fluid according to the United States and the European Pharmacopeias.
  • Used for dissolving materials in test and for rinsing sterile devices or materials
  • Releases contaminating organisms from the device or material in test without affecting their viability
  • Proteose Peptone No. 3 acts as a stabilizer for microorganisms, maintaining their viability
  • Terminally sterilized inside a USP infusion bottle with septum for sterile extraction
  • Sterile bottle packaged in two Stericlin™ bags for use in sterile production areas
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance USP, EP
Content And Storage Store containers in the dark at 5°-25° C
Form Liquid
Product Type Diluting/Rinsing Fluid
Sterility Sterile
Sterilization Method Autoclaved
Without Additives No Polysorbate 80
Format Infusion bottles double wrapped in Stericlin™ bags
Formulation Per Liter Purified Water: Peptic Digest of Animal Tissue - 1 g
For Use With (Application) For dissolving materials and for rinsing devices or materials
pH 7.1 ± 0.2
Quantity 100 mL