Learn More
R&D Systems™ FlowX Human Natural Killer Cell Killing Flow Cytometry Kit
Product Code. 18123591
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
25 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18123591 100 Tests 25 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18123591 Supplier R&D Systems™ Supplier No. FMC032
  • ONLINE EXCLUSIVE PRICE
    676.00€
    639.45€ / 25 tests
    Save 36.55€
    5% Off
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
Flow Cytometry Kit
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Quantity 100 Tests
Species Mouse