Learn More
Tork Floor Stand Dispenser W1
Shop All Tork Products
missing translation for 'orderingAttributeHoverText'
Color:
Red, Smoke
Translucent Blue
missing translation for 'unitSize'
Each
Description
- Ideal for a varied workflow - you can place our mobile dispenser where you need it.
- Cut back on maintenance with our high-capacity stand designed for easy filling and refilling.
- Save precious time by adding a matching bin liner-holder to the stand.
- Compatible with a variety of Tork wiping papers and cleaning cloths, so you can pick the right product for your workspace.
Specifications
Specifications
| Product Type | Floor Stand Wiper Dispenser |
| Color | Red, Smoke |
| Alternate Part Number | 7322540430578 |
| Length (Metric) | 64.6 cm |
| Width (Metric) | 53 cm |
| Height (Metric) | 100.6 cm |
| Holds | Wipes |
| Material | Metal, Plastic |
| Mounting | Freestanding |
| No. of Compartments | 1 |
| Show More |