Learn More
Key Instruments™ Flo Rite™ FR5000-S Model Clear Acrylic Flowmeter with Standard Inlet Valve
Click to view available options
Airflow Rate:
100 to 1400 LPM
100 to 700 LPM
400 to 3000 LPM
Unit Size:
Each
Specifications
Specifications
| Type | Flowmeter |
| Accuracy | ±2% |
| Airflow Rate | 100 to 700 LPM |
| Float Material | Stainless Steel |
| Pressure Rating | 100 psig |
| Connections | 1 in. NPT (F) |
| For Use With (Application) | Air sampling equipment aquaculture, Desalinization equipment, Gas analyzers, Medical systems, Photoprocessing equipment, Water treatment and distribution systems |
| Height (Metric) | 266 mm |
| Internal Part Material | Acrylic |
| Model | FR5000-S |
| Show More |