Learn More
Key Instruments™ Flo Rite™ FR4000 Model Clear Acrylic Flowmeter with Standard Inlet Valve
Product Code. 11978034
Change view
Click to view available options
Airflow Rate:
0.4 to 5 LPM
10 to 100 LPM
2 to 20 LPM
3 to 30 LPM
4 to 50 LPM
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Airflow Rate Unit Size
11978034 2 to 20 LPM Each
11948034 0.4 to 5 LPM Each
11988034 3 to 30 LPM Each
11998034 4 to 50 LPM Each
11908044 10 to 100 LPM Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
Product Code. 11978034 Supplier Key Instruments™ Supplier No. FR4A42BVBN
  • ONLINE EXCLUSIVE PRICE
    114.99€
    109.00€ / Pack of 1
    Save 5.99€
    5% Off
In Stock
Add to basket
Edge
Add to basket
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
Specifications
Type Flowmeter
Accuracy ±3%
Airflow Rate 2 to 20 LPM
Float Material Stainless Steel
Pressure Rating 100 psig
Connections 1/8 in. NPT (F)
For Use With (Application) Air sampling equipment aquaculture, Desalinization equipment, Gas analyzers, Medical systems, Photoprocessing equipment, Water treatment and distribution systems
Height (Metric) 165 mm
Internal Part Material Acrylic
Model FR4000
Repeatability ±0.5%
Width (Metric) 34.7 mm
Scale Full scale
Seal Material Buna-N
Temperature (Metric) 65°C
Valve Standard inlet valve