Learn More
SymLine FlexTube Holder
Click to view available options
For Use With:
FlexTube Tubing
Unit Size:
Each
Description
- Securely clamps FlexTube
- Mountable to flat surface inside cabinet or on a wall
Specifications
Specifications
| Description | Tube holder for Symline FlexTube, suitable for all cabinets, material PA66 halogen-free |
| Material | PTFE-EC |
| For Use With | FlexTube Tubing |
| For Use With (Application) | Clamps Tubing in Place |
| Product Type | Tube Holder |