Learn More
Saint-Gobain Versilon™ R-3400 UV Resistant Acid Transfer Tubing
Product Code. p-8014011
Click to view available options
Diameter (Metric) Inner:
1.6 mm
4.8 mm
6.4 mm
Diameter (Metric) Outer:
3.2 mm
8 mm
9.6 mm
Thickness (Metric):
0.8 mm
1.6 mm
Unit Size:
15m
Each
Product Code. 10616385
Brand: Saint-Gobain AAE00002
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Ideal for virtually any permanent or temporary chemical transfer application, Saint Gobain Performance Plastics™ Versilon™ R-3400 UV Resistant Acid Transfer Tubing combines suppleness and flexibility with resistance to a wide range of chemicals.
Features and Benefits
  • Black in color to protect light-sensitive fluids
  • Compatible with a wide range of chemicals
  • Ozone resistant
  • High temperature rating for excellent burn resistance
  • Low compression set to minimize permanent deformation
Typical Applications
  • Acid and caustic transfer
  • Corrosive diffusable gases
  • Fertilizer and pesticide dispensing
  • Electrical insulation lines
  • Ink and adhesive dispensing
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inner 1.6 mm
Diameter (Metric) Outer 3.2 mm
Thickness (Metric) 0.8 mm
Packaging Quantity Each
Color Black
Flexibility Flexible
Maximum Pressure 4.14 bar