Learn More
De Buyer™ Flexible Spatula
Product Code. 11942625
Click to view available options
Length (Metric):
240 mm
340 mm
420 mm
Unit Size:
Each
Product Code. 11942625
Brand: De Buyer™ 4891.34N
Edge
Scoop, spread and scrape ingredients using the De Buyer™ Flexible Spatula. The spatula can withstand temperatures up to 100°C.
  • Thermoplasticized
  • Up to 100 degrees maximum
  • Plastic handle
Specifications
Quantity 1 Ea.
Product Type Flexible Spatula
Temperature (Metric) Max 100°C
Material Rubber
Length (Metric) 340 mm