Learn More
Copan Italia™ Flexible Inoculation Loop
Product Code. 11766538
Click to view available options
Volume:
10 μL
Unit Size:
Pack of 4000
Product Code. 11766538
Brand: Copan Italia™ 179CS20
Only null left
Add to basket
Add to basket
Used for problem free planting and streaking of cultures.
  • No rough plastic edges, flashing or burrs on the loop head helps problem free planting and streaking of cultures
  • Loops do not cut or gouge the agar surface during streaking
Specifications
Disposable Single-use
Volume 10 μL
Material Flexible soft plastic
Color Light blue
Product Type Inoculating Loop
Quantity 4000