Learn More
Esculape Laboratoire™ Portable First Aid Kit in Red Pouch
Product Code. 11778383
Change view
Click to view available options
Width (Metric):
50 mm
170 mm
Sufficient For:
1 to 4 People
4 to 10 People
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Sufficient For Unit Size
11778383 50 mm 1 to 4 People Each
11718383 170 mm 4 to 10 People Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11778383 Supplier Esculape Laboratoire™ Supplier No. 7240107
  • ONLINE EXCLUSIVE PRICE
    45.06€
    24.22€ / Pack of 1
    Save 20.84€
    46% Off
Estimated Shipment: 12-11-2026
Handle emergency medical treatment with the Esculape™ Portable First Aid Kit in Red Pouch . It includes the essential tools, bandages and burn components to handle immediate care.
  • Hydrogel burn compress helps to reduce swelling While providing immediate cooling, relieves pain and moisturizes the skin.
  • Survival blanket
  • Bandages, compresses,,forceps. gloves
Specifications
Depth (Metric) 50 mm
For Use With (Application) For professional use, sports activities or trips
Length (Metric) 120 mm
Product Type First Aid Kit
Color Red
Includes 1 survival blanket, 2 bags of 5 compresses, 5000 x 20mm 1 roll sticking plaster, 3000 x 70mm 2 gauze bandages, 10 adhesive bandages, 4 fixing clips for bands, 3 alcoholic compresses, 3 compresses to chlorhexidine, 3 anti-blows compresses, 3 mosquito compresses, 1 pair of scissors, 1 splinter tweezers, 2 pairs of vinyl gloves, 1 guide
Sufficient For 1 to 4 People
Width (Metric) 50 mm
Height (Metric) 180 mm