Learn More
Proteintech™ FlexAble CoraLite™ 488 Antibody Labeling Kit for Mouse IgG1
Shop All Proteintech Products
Click to view available options
Quantity:
10 x 0.5 μL Reactions
200 x 0.5 μL Reactions
50 x 0.5 μL Reactions
Unit Size:
10 reactions
200 reactions
50 reactions
Description
FlexAble CoraLite™ 488 Antibody Labeling Kit for Mouse IgG1 is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite™ 488 in any buffer condition to mouse IgG1 primary antibodies from any supplier.
- One labeling reaction requires only 0.5 μg of antibody, regardless of the antibody concentration
- Label your mouse IgG1 antibody in <10 min without any additional equipment
Specifications
Specifications
| Kit Contents | 50 μL CoraLite™ 488 FlexLinker for Mouse IgG1, 100 μL FlexQuencher for Mouse IgG1, 500 μL FlexBuffer, Manual, Protocol Magnet |
| Quantity | 50 x 0.5 μL Reactions |
| Content And Storage | 1 Year at -20°C or 6 Months at +4°C, Avoid Light |
| Excitation/Emission | 500 nm/520 nm |
| Labeling Target | IgG1 |
| Species | Mouse |