Learn More
ZVG™ Fleece All Purpose Cloths
Product Code. 17823505
Click to view available options
Color:
Blue
Green
Pink
Yellow
Unit Size:
Pack of 200
Product Code. 17823505
Brand: ZVG™ 1001000
Only null left
Add to basket
Add to basket
Versatile and durable, this multipurpose cloth is highly absorbent and offers a firm grip. Available in various colors.
Industrial microfiber cloths are suitable for streak-free cleaning of all smooth surfaces (plastic, glass, stainless steel, wood, etc.). With these machine-washable cloths (up to 90°C, without fabric softener), stubborn dirt can often be removed without the use of cleaning agents. Non-woven, all-purpose cloths quickly absorb large quantities of liquids. The universal wipes are suitable for many cleaning jobs in commercial kitchens and restaurants. The zetPutz™ range also includes universal cloths, sponges, and dishcloths for other cleaning tasks.
  • Universally applicable
  • Highly absorbent
  • Durable
  • Stable grip
  • Available in different colors
Specifications
Product Type Cloth
Length (English) 15 in.
Length (Metric) 38 cm
Width (English) 15 in.
Width (Metric) 38 cm
Material Fleece
Color Blue
Disposable Reusable
Packaging Quantity 200 Cloths