Learn More
Flavoxate Hydrochloride, TRC
Product Code. 30397276
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30397276 1 g
30407969 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30397276 Supplier Toronto Research Chemicals Supplier No. TRCF3735001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3717-88-2
Molecular Formula C24 H25 N O4 . Cl H
Molecular Weight (g/mol) 427.92
Synonym 4H-1-Benzopyran-8-carboxylic acid, 3-methyl-4-oxo-2-phenyl-, 2-(1-piperidinyl)ethyl ester, hydrochloride (1:1), 4H-1-Benzopyran-8-carboxylic acid, 3-methyl-4-oxo-2-phenyl-, 2-(1-piperidinyl)ethyl ester, hydrochloride (9CI), 4H-1-Benzopyran-8-carboxylic acid, 3-methyl-4-oxo-2-phenyl-, 2-piperidinoethyl ester, hydrochloride (6CI,7CI,8CI), 1-Piperidineethanol, 3-methyl-4-oxo-2-phenyl-4H-1-benzopyran-8-carboxylate (ester), hydrochloride (8CI), 2-(Piperidin-1-yl)ethyl 3-methyl-4-oxo-2-phenyl-4H-chromene-8-carboxylate hydrochloride, Bladderon, DW 61, Flavoxate hydrochloride, Genurin, NSC 114649, Patricin, Peflate, Piperidinoethyl-3-methylflavone-8-carboxylate hydrochloride, Rec 7/0040, Spasmal, Spasuret, Urispas
IUPAC Name 2-piperidin-1-ylethyl 3-methyl-4-oxo-2-phenylchromene-8-carboxylate;hydrochloride
SMILES Cl.CC1=C(Oc2c(cccc2C1=O)C(=O)OCCN3CCCCC3)c4ccccc4
Specifications
Quantity 1 g
InChI Formula InChI=1S/C24H25NO4.ClH/c1-17-21(26)19-11-8-12-20(23(19)29-22(17)18-9-4-2-5-10-18)24(27)28-16-15-25-13-6-3-7-14-25;/h2,4-5,8-12H,3,6-7,13-16H2,1H3;1H
Formula Weight 427.155
Chemical Name or Material Flavoxate Hydrochloride