Learn More
Flavokawain C, TRC
Specifications
Specifications
| Chemical Name or Material | Flavokavain C |
| CAS | 37308-75-1 |
| Molecular Formula | C17 H16 O5 |
| Quantity | 1 g |
| Synonym | Flavokawain C, 4-Hydroxyflavokavain B |
| InChI Formula | InChI=1S/C17H16O5/c1-21-13-9-15(20)17(16(10-13)22-2)14(19)8-5-11-3-6-12(18)7-4-11/h3-10,18,20H,1-2H3/b8-5+ |
| SMILES | COc1cc(O)c(C(=O)\C=C\c2ccc(O)cc2)c(OC)c1 |
| IUPAC Name | (E)-1-(2-hydroxy-4,6-dimethoxy-phenyl)-3-(4-hydroxyphenyl)prop-2-en-1-one |
| Molecular Weight (g/mol) | 300.3 |
| Formula Weight | 300.0998 |