Learn More
Heidolph™ Flat Rubber Mat for Hei-SHAKE Orbital Core 2000
Shop All Heidolph Instruments Products
Click to view available options
For Use With (Equipment):
Hei-SHAKE Orbital Core 2000
Unit Size:
Each
Description
Non-slip, flat rubber mat to be placed on the integrated shaker's platform and optionally on the perforated platform 2000, e.g. for the multi-tier attachment 2000.
Material: EPDM
Characteristics:
- High resistance to temperatures up to 134°C
- High resistance to acids and alkalis
- Autoclavable
Specifications
Specifications
| Product Type | Non-Slip Mat |
| Material | EPDM |
| For Use With (Equipment) | Hei-SHAKE Orbital Core 2000 |